Learn More
Biomatik Corporation Guinea pig C Reactive Protein (CRP) ELISA Kit
Supplier:  Biomatik Corporation EKU02822
Encompass_Preferred
Guinea pig C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49254. Gene ID: 100727100. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2169