Learn More
Biomatik Corporation Human Calponin 2 (CNN2) ELISA Kit
Supplier:  Biomatik Corporation EKU02891
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.122ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q99439. Target Synonyms: Neutral calponin; Calponin H2, smooth muscle. Restriction: For Research Use Only.
Catalog No. 50-229-2178