Learn More
Biomatik Corporation Human Carbonyl Reductase 3 (CBR3) ELISA Kit
Supplier:  Biomatik Corporation EKU02928
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.060ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O75828. Target Synonyms: SDR21C2; Short Chain Dehydrogenase/Reductase Family 21C,Member 2; NADPH-dependent carbonyl reductase 3. Restriction: For Research Use Only.
Catalog No. 50-229-2182