Learn More
Biomatik Corporation Rat Catechol-O-Methyltransferase (COMT) ELISA Kit
Supplier:  Biomatik Corporation EKU03012
Encompass_Preferred
Rat Catechol-O-Methyltransferase (COMT) ELISA Kit for detecting COMT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22734. Gene ID: 24267. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2197