Learn More
Biomatik Corporation Human Cluster Of Differentiation 160 (CD160) ELISA Kit
Supplier:  Biomatik Corporation EKU03227
Encompass_Preferred
Human Cluster Of Differentiation 160 (CD160) ELISA Kit for detecting CD160 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95971. Gene ID: 11126. Target Synonyms: BY55; NK1; NK28; CD160 Antigen; Natural killer cell receptor BY55. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2229