Learn More
Biomatik Corporation Human Coiled Coil Domain Containing Protein 3 (CCDC3) ELISA Kit
Supplier:  Biomatik Corporation EKU03292
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.116ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9BQI4. Target Synonyms: Favine; Fat/Vessel-Derived Secretory Protein. Restriction: For Research Use Only.
Catalog No. 50-229-2243