Learn More
Biomatik Corporation Rat Complement Factor H (CFH) ELISA Kit
Supplier:  Biomatik Corporation EKU03425
Encompass_Preferred
Rat Complement Factor H (CFH) ELISA Kit for detecting CFH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.77ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: G3V9R2. Gene ID: 155012. Target Synonyms: CF-H; FH; FHL1; ARMD4; ARMS1; CFHL3; HF1; HF2; HUS; H Factor 2; Age-Related Maculopathy Susceptibility 1; Adrenomedullin binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2255