Learn More
Biomatik Corporation Mouse Cross Linked N-Telopeptide Of Type I Collagen (NTXI) ELISA Kit
Supplier:  Biomatik Corporation EKU03510
Encompass_Preferred
Sample Type: Serum, plasma, urine and other biological fluids. Species Reactivity: Mouse. Detection Range: 2.47-200ng/mL. Sensitivity: 0.93ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Competitive. Assay Time: 2 hours. UniProt ID: Not Available. Target Synonyms: NTX-I; NTX1. Restriction: For Research Use Only.
Catalog No. 50-229-2265