Learn More
Biomatik Corporation Rat Cyclophilin B (CYPB) ELISA Kit
Supplier:  Biomatik Corporation EKU03554
Encompass_Preferred
Rat Cyclophilin B (CYPB) ELISA Kit for detecting CYPB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24368. Gene ID: 64367. Target Synonyms: PPIB; PPI-B; CYP-S1; CyP-B; SCYLP; PPIase B; Rotamase B; S-cyclophilin; Peptidylprolyl Isomerase B; Peptidyl-Prolyl Cis-Trans Isomerase B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2275