Learn More
Biomatik Corporation Rat Cystathionine (CTH) ELISA Kit
Supplier:  Biomatik Corporation EKU03558
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.053ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P18757. Target Synonyms: CSE; Cystathionase; Cysteine-protein sulfhydrase; Gamma-cystathionase. Restriction: For Research Use Only.
Catalog No. 50-229-2277