Learn More
Biomatik Corporation Human Cysteine And Glycine Rich Protein 1 (CSRP1) ELISA Kit
Supplier:  Biomatik Corporation EKU03570
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P21291. Target Synonyms: CRP1; CRP; CSRP; CYRP; HEL-141; Cysteine-rich protein 1; Epididymis luminal protein 141. Restriction: For Research Use Only.
Catalog No. 50-229-2279