Learn More
Biomatik Corporation Human Cytochrome P450 1A2 (CYP1A2) ELISA Kit
Supplier:  Biomatik Corporation EKU03593
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.625-40ng/mL. Sensitivity: 0.257ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P05177. Target Synonyms: CP12; P3-450; P450(PA); Cytochrome P450,Family 1,Subfamily A,Polypeptide 2. Restriction: For Research Use Only.
Catalog No. 50-229-2283