Learn More
Biomatik Corporation Human Cytochrome P450 24A1 (CYP24A1) ELISA Kit
Supplier:  Biomatik Corporation EKU03596
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q07973. Target Synonyms: CP24; CYP24; P450-CC24; Vitamin D 24-Hydroxylase; 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial ; Cytochrome P450 Family 24 Subfamily A Polypeptide 1. Restriction: For Research Use Only.
Catalog No. 50-229-2285