Learn More
Biomatik Corporation Human Defensin Alpha 1B (DEFa1B) ELISA Kit
Supplier:  Biomatik Corporation EKU03649
Encompass_Preferred
Human Defensin Alpha 1B (DEFa1B) ELISA Kit for detecting DEFa1B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P59665. Gene ID: 1667. Target Synonyms: HNP-1; DEFA2; MRS; Neutrophil defensin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2294