Learn More
Biomatik Corporation Human Defensin Beta 131 (DEFb131) ELISA Kit
Supplier:  Biomatik Corporation EKU03666
Encompass_Preferred
Human Defensin Beta 131 (DEFb131) ELISA Kit for detecting DEFb131 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P59861. Gene ID: 644414. Target Synonyms: DEFB-31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2296