Learn More
Biomatik Corporation Mouse Deoxyribonuclease I Like Protein 3 (DNASE1L3) ELISA Kit
Supplier:  Biomatik Corporation EKU03693
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O55070. Target Synonyms: DHP2; DNAS1L3; LSD; Deoxyribonuclease Gamma; Liver and spleen DNase; DNase I homolog protein DHP2. Restriction: For Research Use Only.
Catalog No. 50-229-2303