Learn More
Biomatik Corporation Human Dopa Decarboxylase (DDC) ELISA Kit
Supplier:  Biomatik Corporation EKU03777
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.61ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P20711. Target Synonyms: AADC; Aromatic L-Amino Acid Decarboxylase; DOPA decarboxylase. Restriction: For Research Use Only.
Catalog No. 50-229-2318