Learn More
Biomatik Corporation Rat Dopamine Receptor D1 (DRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU03782
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.058ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P18901. Target Synonyms: DR-D1; D1R; DADR; DRD1A. Restriction: For Research Use Only.
Catalog No. 50-229-2320