Learn More
Biomatik Corporation Rat Doublecortin (DCX) ELISA Kit
Supplier:  Biomatik Corporation EKU03791
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: DBCN; DC; LISX; SCLH; XLIS; Doublin; Doublecortex; Lissencephalin-X; Lissencephaly,X-Linked; Neuronal migration protein doublecortin. Restriction: For Research Use Only.
Catalog No. 50-229-2323