Learn More
Biomatik Corporation Mouse Dual Specificity Tyrosine Phosphorylation Regulated Kinase 1A (DYRK1A) ELISA Kit
Supplier:  Biomatik Corporation EKU03805
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: DYRK; DYRK1; HP86; MNB; MNBH; Dual specificity YAK1-related kinase; Protein kinase minibrain homolog. Restriction: For Research Use Only.
Catalog No. 50-229-2325