Learn More
Biomatik Corporation Rat ELAV Like Protein 1 (ELAVL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03858
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: ELAV1; HUR; Hua; MelG; Hu Antigen R; Embryonic Lethal,Abnormal Vision Like 1. Restriction: For Research Use Only.
Catalog No. 50-229-2336