Learn More
Biomatik Corporation Mouse Endoglin (ENG) ELISA Kit
Supplier:  Biomatik Corporation EKU03873
Encompass_Preferred
Mouse Endoglin (ENG) ELISA Kit for detecting ENG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63961. Gene ID: 13805. Target Synonyms: CD105; END; HHT1; ORW; ORW1; Osler-Rendu-Weber Syndrome 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2338