Learn More
Biomatik Corporation Rat Enolase 1 (ENO1) ELISA Kit
Supplier:  Biomatik Corporation EKU03908
Encompass_Preferred
Rat Enolase 1 (ENO1) ELISA Kit for detecting ENO1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04764. Gene ID: 24333. Target Synonyms: NNE; ENO1L1; MPB1; PPH; Alpha Enolase; Enolase 1; Phosphopyruvate hydratase; Plasminogen-binding protein; 2-phospho-D-glycerate hydro-lyase; C-myc promoter-binding. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2343