Learn More
Biomatik Corporation Mouse Epstein Barr Virus Induced Protein 3 (EBI3) ELISA Kit
Supplier:  Biomatik Corporation EKU03956
Encompass_Preferred
Mouse Epstein Barr Virus Induced Protein 3 (EBI3) ELISA Kit for detecting EBI3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35228. Gene ID: 50498. Target Synonyms: IL27B; IL27-B; Interleukin 27B; Epstein-Barr virus-induced gene 3 protein; EBV-induced gene 3 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2354