Learn More
Biomatik Corporation Human Extracellular Matrix Protein 1 (ECM1) ELISA Kit
Supplier:  Biomatik Corporation EKU04004
Encompass_Preferred
Human Extracellular Matrix Protein 1 (ECM1) ELISA Kit for detecting ECM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40pg/ml. Sensitivity: 0.239pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16610. Gene ID: 1893. Target Synonyms: Secretory component p85. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2364