Learn More
Biomatik Corporation Human Fatty Acid Desaturase 1 (FADS1) ELISA Kit
Supplier:  Biomatik Corporation EKU04047
Encompass_Preferred
Human Fatty Acid Desaturase 1 (FADS1) ELISA Kit for detecting FADS1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60427. Gene ID: 3992. Target Synonyms: D5D; FADS6; FADSD5; LLCDL1; TU12; Delta-5 Desaturase; Delta (5) fatty acid desaturase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2368