Learn More
Biomatik Corporation Human Ferrochelatase (FECH) ELISA Kit
Supplier:  Biomatik Corporation EKU04077
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P22830. Target Synonyms: EPP; FCE; Protoheme Ferrolyase; Protoporphyria; Heme synthase; Protoheme ferro-lyase. Restriction: For Research Use Only.
Catalog No. 50-229-2370