Learn More
Biomatik Corporation Rat Fibrinogen Like Protein 2 (FGL2) ELISA Kit
Supplier:  Biomatik Corporation EKU04111
Encompass_Preferred
Rat Fibrinogen Like Protein 2 (FGL2) ELISA Kit for detecting FGL2 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.12-5000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q32Q89. Target Synonyms: T49; PT49; Fibroleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2376