Learn More
Biomatik Corporation Human Fibroblast Growth Factor 17 (FGF17) ELISA Kit
Supplier:  Biomatik Corporation EKU04141
Encompass_Preferred
Human Fibroblast Growth Factor 17 (FGF17) ELISA Kit for detecting FGF17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60258. Gene ID: 8822. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2380