Learn More
Biomatik Corporation Human Fibrous Sheath Interacting Protein 1 (FSIP1) ELISA Kit
Supplier:  Biomatik Corporation EKU04206
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.118ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8NA03. Target Synonyms: HDS10. Restriction: For Research Use Only.
Catalog No. 50-229-2387