Learn More
Biomatik Corporation Human FK506 Binding Protein 4 (FKBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU04232
Encompass_Preferred
Human FK506 Binding Protein 4 (FKBP4) ELISA Kit for detecting FKBP4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02790. Gene ID: 2288. Target Synonyms: FKBP52; FKBP59; HBI; Hsp56; p52; Rotamase; 51 kDa FK506-binding protein; 59 kDa immunophilin; HSP-binding immunophilin; Peptidyl-prolyl cis-trans isomerase FKBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2391