Learn More
Biomatik Corporation Human Flightless I Homolog (FLII) ELISA Kit
Supplier:  Biomatik Corporation EKU04237
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 78.12-5,000pg/mL. Sensitivity: 29.2pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q13045. Target Synonyms: FLI; FLIL; Fli1. Restriction: For Research Use Only.
Catalog No. 50-229-2393