Learn More
Biomatik Corporation Mouse Follistatin (FS) ELISA Kit
Supplier:  Biomatik Corporation EKU04254
Encompass_Preferred
Mouse Follistatin (FS) ELISA Kit for detecting FS in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.91ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P47931. Gene ID: 14313. Target Synonyms: FST; Activin-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2395