Learn More
Biomatik Corporation Rat Forkhead Box Protein O1 (FOXO1) ELISA Kit
Supplier:  Biomatik Corporation EKU04260
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.122ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: FOX-O1; FKH1; FKHR; FOXO1A; Forkhead Homolog In Rhabdomyosarcoma. Restriction: For Research Use Only.
Catalog No. 50-229-2397