Learn More
Biomatik Corporation Human G Protein Coupled Receptor 35 (GPR35) ELISA Kit
Supplier:  Biomatik Corporation EKU04303
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9HC97. Target Synonyms: KYNA receptor; Kynurenic acid receptor. Restriction: For Research Use Only.
Catalog No. 50-229-2409