Learn More
Biomatik Corporation Human G Protein Coupled Receptor, Family C, Group 5, Member A (GPRC5A) ELISA Kit
Supplier:  Biomatik Corporation EKU04306
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8NFJ5. Target Synonyms: RAIG1; RAI3, GPCR5A; Retinoic Acid Induced 3; Orphan G-protein-coupling receptor PEIG-1; Retinoic acid-induced gene 1 protein. Restriction: For Research Use Only.
Catalog No. 50-229-2411