Learn More
Biomatik Corporation Human Gastric Intrinsic Factor (GIF) ELISA Kit
Supplier:  Biomatik Corporation EKU04343
Encompass_Preferred
Human Gastric Intrinsic Factor (GIF) ELISA Kit for detecting GIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 51.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P27352. Gene ID: 2694. Target Synonyms: TCN3; IF; IFMH; INF; Transcobalamin III; Vitamin B Synthesis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2420