Learn More
Biomatik Corporation Mouse Glucocorticoid Receptor Alpha (GRa) ELISA Kit
Supplier:  Biomatik Corporation EKU04397
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.121ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P06537. Target Synonyms: GR-A. Restriction: For Research Use Only.
Catalog No. 50-229-2426