Learn More
Biomatik Corporation Mouse Glucose 6 Phosphate Isomerase (GPI) ELISA Kit
Supplier:  Biomatik Corporation EKU04410
Encompass_Preferred
Mouse Glucose 6 Phosphate Isomerase (GPI) ELISA Kit for detecting GPI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06745. Gene ID: 14751. Target Synonyms: AMF; NLK; PHI; SA36; Phosphoglucose Isomerase; Autocrine motility factor; Neuroleukin; Phosphohexose isomerase; Sperm antigen 36. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2430