Learn More
Biomatik Corporation Rat Glucose Transporter 1 (GLUT1) ELISA Kit
Supplier:  Biomatik Corporation EKU04414
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 1.25-80ng/mL. Sensitivity: 0.51ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC2A1; SLC2-A1; GLUT; Solute Carrier Family 2 Member 1,Facilitated Glucose Transporter; Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter. Restriction: For Research Use Only.
Catalog No. 50-229-2431