Learn More
Biomatik Corporation Human Glutamate Decarboxylase 1, Brain (GAD1) ELISA Kit
Supplier:  Biomatik Corporation EKU04437
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GAD67; SCP; 67 kDa glutamic acid decarboxylase; Glutamate decarboxylase 67 kDa isoform. Restriction: For Research Use Only.
Catalog No. 50-229-2435