Learn More
Biomatik Corporation Mouse Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3) ELISA Kit
Supplier:  Biomatik Corporation EKU04451
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GLUR-K3; GLUR3; GLURC; gluR-C; AMPA-selective glutamate receptor 3. Restriction: For Research Use Only.
Catalog No. 50-229-2437