Learn More
Biomatik Corporation Mouse Glutathione Reductase (GR) ELISA Kit
Supplier:  Biomatik Corporation EKU04484
Encompass_Preferred
Mouse Glutathione Reductase (GR) ELISA Kit for detecting GR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47791. Gene ID: 14782. Target Synonyms: GSR; GLUR; GRD1; Glutathione reductase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2444