Learn More
Biomatik Corporation Human Glycerol-3-Phosphate Acyltransferase, Mitochondrial (GPAM) ELISA Kit
Supplier:  Biomatik Corporation EKU04506
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.55ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9HCL2. Target Synonyms: GPAT1. Restriction: For Research Use Only.
Catalog No. 50-229-2449