Learn More
Biomatik Corporation Mouse Granulin (GRN) ELISA Kit
Supplier:  Biomatik Corporation EKU04556
Encompass_Preferred
Mouse Granulin (GRN) ELISA Kit for detecting GRN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28798. Gene ID: 14824. Target Synonyms: GEP; GP88; PCDGF; PEPI; PGRN; Proepithelin; Acrogranin; Glycoprotein of 88 Kda; Paragranulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2459