Learn More
Biomatik Corporation Mouse Growth Differentiation Factor 1 (GDF1) ELISA Kit
Supplier:  Biomatik Corporation EKU04583
Encompass_Preferred
Mouse Growth Differentiation Factor 1 (GDF1) ELISA Kit for detecting GDF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20863. Gene ID: 14559. Target Synonyms: Embryonic growth/differentiation factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2465