Learn More
Biomatik Corporation Human Growth Differentiation Factor 6 (GDF6) ELISA Kit
Supplier:  Biomatik Corporation EKU04600
Encompass_Preferred
Human Growth Differentiation Factor 6 (GDF6) ELISA Kit for detecting GDF6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6KF10. Gene ID: 392255. Target Synonyms: BMP13; CDMP2; GDF16; Bone morphogenetic protein 13; Growth/differentiation factor 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2470