Learn More
Biomatik Corporation Rat Heat Shock 70kDa Protein 5 (HSPA5) ELISA Kit
Supplier:  Biomatik Corporation EKU04662
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.781-50ng/mL. Sensitivity: 0.32ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: MIF2; BIP; GRP78; Immunoglobulin heavy chain-binding protein; Glucose Regulated Protein 78; Binding Immunoglobulin Protein; Endoplasmic reticulum lumenal Ca binding grp78. Restriction: For Research Use Only.
Catalog No. 50-229-2477