Learn More
Biomatik Corporation Human Hemoglobin Gamma 1 (HBg1) ELISA Kit
Supplier:  Biomatik Corporation EKU04722
Encompass_Preferred
Human Hemoglobin Gamma 1 (HBg1) ELISA Kit for detecting HBg1 in plasma, erythrocyte lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 370.37-30,000ng/ml. Sensitivity: 129.4ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P69891. Gene ID: 3047. Target Synonyms: HB-G1; HBGA; HBGR; HSGGL1; Gamma-1-globin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2484