Learn More
Biomatik Corporation Mouse Hepatoma Derived Growth Factor (HDGF) ELISA Kit
Supplier:  Biomatik Corporation EKU04761
Encompass_Preferred
Mouse Hepatoma Derived Growth Factor (HDGF) ELISA Kit for detecting HDGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51859. Gene ID: 15191. Target Synonyms: HMG1L2; High-Mobility Group Protein 1-Like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2489